EMAIL THIS PAGE TO A FRIEND
Phytotherapy research : PTR
Kavalactone metabolism in the isolated perfused rat liver.
PMID 22407838
Abstract
Metabolic pathways for kavalactone metabolism in humans and rats have been identified, but more detailed description of the enzyme kinetics involved is lacking. The disposition profiles of three of the six major kavalactones (kavain, methysticin and desmethoxyyangonin) and their respective metabolites (p-hydroxykavain, m,p-dihydroxykavain and p-hydroxy-5,6-dehydrokavain) were examined in the perfusate and bile of the isolated perfused rat liver. The metabolism of the kavalactones is first-order in nature with similar decay half-lives. p-Hydroxykavain and m,p-dihydroxykavain were the only metabolites detected in the perfusate. Kavalactone biliary excretion was negligible.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
59780
DL-Kavain, analytical standard
C14H14O3
80488
R-(+)-Methysticin, analytical standard
C15H14O5