EMAIL THIS PAGE TO A FRIEND
BMC research notes
Paclobutrazol treatment as a potential strategy for higher seed and oil yield in field-grown Camelina sativa L. Crantz.
PMID 22410213
Abstract
Camelina (Camelina sativa L. Crantz) is a non-food oilseed crop which holds promise as an alternative biofuel energy resource. Its ability to grow in a variety of climatic and soil conditions and minimal requirements of agronomical inputs than other oilseed crops makes it economically viable for advanced biofuel production. We designed a study to investigate the effect of paclobutrazol [2RS, 3RS)-1-(4-Chlorophenyl)-4,4-dimethyl-2-(1H-1,2,4-triazol-1-yl)pentan-3-ol] (PBZ), a popular plant growth regulator, on the seed and oil yield of Camelina sativa (cv. Celine). A field-based micro-trial setup was established in a randomized block design and the study was performed twice within a span of five months (October 2010 to February 2011) and five different PBZ treatments (Control: T0; 25 mg l-1: T1; 50 mg l-1: T2; 75 mg l-1: T3; 100 mg l-1: T4; 125 mg l-1: T5) were applied (soil application) at the time of initiation of flowering. PBZ at 100 mg l-1 concentration (T4) resulted in highest seed and oil yield by 80% and 15%, respectively. The seed yield increment was mainly due to enhanced number of siliques per plant when compared to control. The PBZ - treated plants displayed better photosynthetic leaf gas exchange characteristics, higher chlorophyll contents and possessed dark green leaves which were photosynthetically active for a longer period and facilitated higher photoassimilation. We report for the first time that application of optimized PBZ dose can be a potential strategy to achieve higher seed and oil yield from Camelina sativa that holds great promise as a biofuel crop in future.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46046
Paclobutrazol, PESTANAL®, analytical standard
C15H20ClN3O