EMAIL THIS PAGE TO A FRIEND
Cardiovascular research
Ablation of junctin or triadin is associated with increased cardiac injury following ischaemia/reperfusion.
PMID 22411973
Abstract
Junctin and triadin are calsequestrin-binding proteins that regulate sarcoplasmic reticulum (SR) Ca(2+) release by interacting with the ryanodine receptor. The levels of these proteins are significantly down-regulated in failing human hearts. However, the significance of such decreases is currently unknown. Here, we addressed the functional role of these accessory proteins in the heart's responses to ischaemia/reperfusion (I/R) injury. Isolated mouse hearts were subjected to global I/R, and contractile parameters were assessed in wild-type (WT), junctin-knockout (JKO), and triadin-knockout (TKO) hearts. Both JKO and TKO were associated with significantly depressed post-I/R contractile recovery. However, ablation of triadin resulted in the most severe post-I/R phenotype. The additional contractile impairment of TKO hearts was not related to a mitochondrial death pathway, but attributed to endoplasmic reticulum (ER) stress-mediated apoptosis. Activation of the X-box-binding protein-1 and transcriptional up-regulation of C/EBP-homologous protein (CHOP) provided a molecular mechanism of caspase-12-dependent apoptosis in myocytes. In addition, elevation of cytosolic Ca(2+) during reperfusion was associated with the activation of calpain proteases and troponin I breakdown. Accordingly, treatment with the calpain inhibitor MDL-28170 significantly ameliorated post-I/R impairment of contractile recovery in intact hearts. These findings indicate that deficiency of either junctin or triadin impairs the contractile recovery in post-ischaemic hearts, which appears to be primarily attributed to increased ER stress and activation of calpain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M6690
MDL 28170, ≥95% (HPLC)
C22H26N2O4