EMAIL THIS PAGE TO A FRIEND
Masui. The Japanese journal of anesthesiology
[Anesthetic management of a patient with pulmonary hypertension due to a large anterior mediastinal tumor anticipated by preoperative computed tomography].
PMID 22413448
Abstract
A 74-year-old man underwent an operation for an extremely large mediastinal tumor. Despite the lack of respiratory difficulty or distention of the jugular veins, preoperative computed tomography showed suppression of the left atrium by the huge tumor. We suspected pulmonary hypertension and monitored continuously with a pulmonary catheter. The Pp/Ps ratio decreased gradually from 0.8 to 0.7 by continuous administration of alprostadil or olprinone. After tumor resection, the Pp/Ps ratio was reduced and maintained at 0.2 even after alprostadil and olprinone administration was discontinued. The tumor was considered to have caused the severe pulmonary hypertension, as anticipated from the preoperative CT.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O7389
Olprinone hydrochloride, ≥98% (HPLC), solid
C14H10N4O · HCl