EMAIL THIS PAGE TO A FRIEND
Applied Immunohistochemistry & Molecular Morphology
Immunohistochemical detection of glypican-5 in paraffin-embedded material: an optimized method for a novel research antibody.
PMID 22415220