EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Assessment of the levels of persistent organic pollutants and 1-hydroxypyrene in blood and urine samples from Mexican children living in an endemic malaria area in Mexico.
PMID 22415648
Abstract
The aim of this study was to evaluate the exposure levels to persistent organic pollutants and 1-hydroxypyrene in children living in an endemic malaria zone in Mexico. The blood levels for 1,1,1-trichloro-2,2-bis(p-chlorophenyl)ethane (DDT), 1,1-dichloro-2,2-bis(p-chlorophenyl)ethylene (DDE) and lindane ranged from 15.4 to 17,886.5 ng/g lipid, 6,624.3 to 100,119.0 ng/g lipid, and 351.1 to 6,153.8 ng/g lipid, respectively. For total polychlorinated biphenyls the blood levels ranged from 2,584.9 to 14,547.9 ng/g lipid. Regarding urinary 1-hydroxypyrene levels, the mean level was 2.9 ± 3.1 μmol/mol creatinine. In conclusion, the children in our study are exposed to levels higher than normal to mixtures of environmental contaminants.