EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Development of a rapid method for the determination and confirmation of nitroimidazoles in six matrices by fast liquid chromatography-tandem mass spectrometry.
PMID 22417613
Abstract
A rapid liquid chromatography-tandem mass spectrometric (LC-MS/MS) method was developed to identify and to quantify nitroimidazoles, metronidazole (MNZ), ronidazole (RNZ) and dimetridazole (DMZ) and their corresponding hydroxy metabolites, MNZ-OH and 2-hydroxymethyl-1-methyl-5-nitroimidazole (HMNNI) in plasma, milk, muscle, egg, honey and feed samples. The same sample clean-up procedure including a novel solid-phase extraction (SPE) on polymeric Strata-SDB cartridges was used for each matrix. The analytes were separated on Kinetex XB C-18 core-shell type HPLC column using isocratic elution mode with a mobile phase containing 0.1% formic acid in water/methanol (88/12, v/v, pH 2.6) at a flow rate of 0.7 ml/min. The main advantage of the developed method is that the analysis time of only 3 min, which is about three to ten times shorter than in other reported HPLC methods. The developed method was validated using a matrix-comprehensive in-house validation strategy. The matrix effect of LC-MS/MS analysis was also investigated. Results are presented from the successful application of the developed method to an incurred pork meat certified reference material and to incur porcine plasmas in a proficiency test in year 2011.
Related Materials