EMAIL THIS PAGE TO A FRIEND
Zhejiang da xue xue bao. Yi xue ban = Journal of Zhejiang University. Medical sciences
[Screening and identifying hepatotoxic components in Aucklandiae Radix with GC-MS].
PMID 22419462
Abstract
To screen the potential hepatotoxic components in Chinese herb medicine Aucklandiae Radix. The potential hepatotoxic components were screened using HepG2 cells labeled with fluorescein diacetate from 25 fractions of Aucklandiae Radix, in which the hepatotoxic compounds were further identified with GC-MS. Ten potential hepatotoxic fractions were screened. The identification results by GC-MS indicated that the main compounds in C09 were dehydrocostuslactone, santamarine (or magnolialide) and reynosin, and in C11 were α-costol and elemol. Dehydrocostuslactone, santamarine (or magnolialide), reynosin, α-costol and elemol are potential hepatotoxic compounds in Aucklandiae Radix.