Hepatology (Baltimore, Md.)
Structural analysis of oval-cell-mediated liver regeneration in rats.
PMID 22419534
We have analyzed the architectural aspects of progenitor-cell-driven regenerative growth in rat liver by applying the 2-acetaminofluorene/partial hepatectomy experimental model. The regeneration is initiated by the proliferation of so-called oval cells. The oval cells at the proximal tips of the ductules have a more differentiated phenotype and higher proliferative rate. This preferential growth results in the formation of a seemingly random collection of small hepatocytes, called foci. These foci have no clonal origin, but possess a highly organized structure, which shows similarities to normal hepatic parenchyma. Therefore, they can easily remodel into the lobular structure. Eventually, the regenerated liver is constructed by enlarged hepatic lobules; no new lobules are formed during this process. The foci of the Solt-Farber experimental hepatocarcinogenesis model have identical morphological features; accordingly, they also represent only regenerative, not neoplastic, growth. Progenitor-cell-driven liver regeneration is a well-designed, highly organized tissue reaction, and better comprehension of the architectural events may help us to recognize this process and understand its role in physiological and pathological reactions.