EMAIL THIS PAGE TO A FRIEND
Glia
Aquaporin-4 regulates extracellular space volume dynamics during high-frequency synaptic stimulation: a gene deletion study in mouse hippocampus.
PMID 22419561
Abstract
Little is known about the physiological roles of aquaporin-4 (AQP4) in the central nervous system. AQP4 water channels are concentrated in endfeet membranes of astrocytes but also localize to the fine astrocytic processes that abut central synapses. Based on its pattern of expression, we predicted that AQP4 could be involved in controlling water fluxes and changes in extracellular space (ECS) volume that are associated with activation of excitatory pathways. Here, we show that deletion of Aqp4 accentuated the shrinkage of the ECS that occurred in the mouse hippocampal CA1 region during activation of Schaffer collateral/commissural fibers. This effect was found in the stratum radiatum (where perisynaptic astrocytic processes abound) but not in the pyramidal cell layer (where astrocytic processes constitute but a minor volume fraction). For both genotypes the ECS shrinkage was most pronounced in the pyramidal cell layer. Our data attribute a physiological role to AQP4 and indicate that this water channel regulates extracellular volume dynamics in the mammalian brain.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
87708
Tetramethylammonium bromide, for ion pair chromatography, ≥99.0% (AT)
C4H12BrN
426296
Tetramethylammonium bromide, ACS reagent, ≥98.0%
C4H12BrN
195758
Tetramethylammonium bromide, 98%
C4H12BrN
87709
Tetramethylammonium bromide, for electrochemical analysis, ≥99.0%
C4H12BrN
87718
Tetramethylammonium chloride, BioUltra, for molecular biology, ≥99.0% (AT)
C4H12ClN
74202
Tetramethylammonium chloride, for ion pair chromatography, ≥99.0% (AT)
C4H12ClN
T19526
Tetramethylammonium chloride, reagent grade, ≥98%
C4H12ClN
T3411
Tetramethylammonium chloride solution, for molecular biology
459135
Tetramethylammonium fluoride, 97%
C4H12FN
T7505
Tetramethylammonium hydroxide pentahydrate, ≥97%
C4H13NO · 5H2O
87741
Tetramethylammonium hydroxide pentahydrate, ≥95.0% (T)
C4H13NO · 5H2O
426318
Tetramethylammonium hydroxide solution, ACS reagent
C4H13NO
334901
Tetramethylammonium hydroxide solution, 25 wt. % in methanol
C4H13NO
331635
Tetramethylammonium hydroxide solution, 25 wt. % in H2O
C4H13NO
328251
Tetramethylammonium hydroxide solution, 10 wt. % in H2O
C4H13NO
235946
Tetramethylammonium iodide, 99%
C4H12IN
425257
Tetramethylammonium nitrate, 95%
C4H12N2O3
02799
Tetramethylammonium sulfate, for ion pair chromatography, ≥99.0% (T)
C8H24N2O4S
87743
Tetramethylammonium sulfate hydrate
C8H24N2O4S · xH2O