EMAIL THIS PAGE TO A FRIEND
Organic letters
Highly efficient synthesis of N-substituted isoindolinones and phthalazinones using Pt nanowires as catalysts.
PMID 22420427
Abstract
A series of N-substituted isoindolinones have been successfully synthesized through the reductive C-N coupling and intramolecular amidation of 2-carboxybenzaldehyde and amines. This one-pot synthesis gives excellent yields using ultrathin Pt nanowires as catalysts under 1 bar of hydrogen. These unsupported catalysts can also be used for the synthesis of phthalazinones in high yield when hydrazine or phenyl hydrazine is used instead of amines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
116017
2-Carboxybenzaldehyde, 97%
C8H6O3