EMAIL THIS PAGE TO A FRIEND
Experimental and clinical endocrinology & diabetes : official journal, German Society of Endocrinology [and] German Diabetes Association
Seronegative myasthenia gravis and Graves' disease. Is there a link?
PMID 22421982
Abstract
Graves' disease (GD) and myasthenia gravis (MG) are common autoimmune diseases but their coexistence is very rare. They may possibly share the same pathogenetic mechanisms. Recent research has shown the involvement of autoantibodies, lymphocytes, cytokines and chemokines in the pathogenesis of MG and GD. It appears that Th17 cell lineage is involved in autoimmune thyroid disease (AITD) and seems to be key factor in the development of both MG and GD.A 34-year-old male with seronegative myasthenia gravis due to thymic hyperplasia was diagnosed with also GD and opthalmopathy. Several diagnostic and therapeutic issues regarding the relevant literature are discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0132
Lanreotide acetate, ≥98% (HPLC)
C54H69N11O10S2 · xC2H4O2