EMAIL THIS PAGE TO A FRIEND
Human reproduction (Oxford, England)
GnRH-agonist versus GnRH-antagonist IVF cycles: is the reproductive outcome affected by the incidence of progesterone elevation on the day of HCG triggering? A randomized prospective study.
PMID 22422777
Abstract
In view of the current debate concerning possible differences in efficacy between the two GnRH analogues used in IVF stimulated cycles, the current study aimed to explore whether progesterone control in the late follicular phase differs when GnRH antagonist is used as compared with GnRH agonist, and if so, to what extent the progesterone rise affects the probability of pregnancy. Overall 190 patients were randomized: 94 in the GnRH-agonist group and 96 in the GnRH-antagonist group. The GnRH-agonist long protocol started on Day 21 of the preceding cycle with intranasal buserelin (600 mg per day). The GnRH-antagonist protocol started on Day 6 of the stimulation with ganirelix or cetrorelix (each 0.25 mg). All blood samples were analysed with the Elecsys analyzer. An intention-to-treat analysis was applied. A progesterone rise >1.5 ng/ml was noticed in 23.0% of the antagonist group, comparable with 24.1% incidence within the agonist group. Per patient randomized, delivery rates were also comparable: 28.1% in the antagonist group and 24.5% in the agonist group (odds ratio = 1.21, 95% confidence interval: 0.63-2.31, P= 0.56). However, there was a reduction in delivery rates when progesterone exceeded the threshold of 1.5 ng/ml, both in the agonist group (9.5 versus 31.8%, P= 0.03) and in the antagonist group (14.3 versus 34.3%, P= 0.07). Although the incidence of a progesterone rise was similar between the two analogues, our findings reconfirm previous observations that insufficient progesterone control (>1.5 ng/ml) on the day of ovulation triggering is related to poor delivery rates in both protocols. The current study has shown that the reproductive outcomes with the two GnRH analogues are comparable. Possible modes of action to circumvent late follicular progesterone rise should be explored. NCT01191710.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B3303 Buserelin acetate salt, ≥90% (HPLC)
C60H86N16O13 · C2H4O2
SML0241 Ganirelix acetate salt, ≥95% (HPLC)
C80H113ClN18O13 · 2C2H4O2