EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Toxicology. Heart-stopping revelation about how Chinese mushroom kills.
PMID 22422955
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G6503
4-Guanidinobutyric acid, ≥98%
C5H11N3O2