EMAIL THIS PAGE TO A FRIEND
Psychosomatics
Wilson's disease--treatment of psychiatric manifestations in pregnancy.
PMID 22424165
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90460
Triethylenetetramine, ≥97.0% (T)
C6H18N4
132098
Triethylenetetramine, technical grade, 60%
C6H18N4