EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Copper(I) mediated facile synthesis of potent tubulin polymerization inhibitor, 9-amino-α-noscapine from natural α-noscapine.
PMID 22425569
Abstract
Facile synthesis of natural α-noscapine analogue, 9-amino-α-noscapine, a potent inhibitor of tubulin polymerization for cancer therapy, is achieved via copper(I) iodide mediated in situ aromatic azidation and reduction of 9-bromo-α-noscapine (obtained by bromination of natural α-noscapine) with NaN(3) in DMSO at 130°C in the presence of L-proline as an amino acid promoter. The protocol developed here avoided isolation of 9-azido-α-noscapine and did not cleave the sensitive C-C bond between two heterocyclic phthalide and isoquinoline units.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
363960
(S,R)-Noscapine, 97%
C22H23NO7