EMAIL THIS PAGE TO A FRIEND
Methods in molecular biology (Clifton, N.J.)
Molecular modeling of lipase binding to a substrate-water interface.
PMID 22426727
Abstract
Interactions of lipases with hydrophobic substrate-water interfaces are of great interest to design improved lipase variants and engineer reaction conditions. This chapter describes the necessary steps to carry out molecular dynamics simulations of Candida antarctica lipase B at tributyrin-water interface using the GROMACS simulation software. Special attention is drawn to the preparation of the protein and the substrate-water interface and to the analysis of the obtained trajectory.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T8626
Glyceryl tributyrate, ≥99%
C15H26O6
91010
Glyceryl tributyrate, puriss., ≥98.5% (GC)
C15H26O6