EMAIL THIS PAGE TO A FRIEND
Lipids
Dietary supplementation with 22-S-hydroxycholesterol to rats reduces body weight gain and the accumulation of liver triacylglycerol.
PMID 22426832
Abstract
This study explores the pharmacokinetics of 22-S-hydroxycholesterol (22SHC) in vivo in rats. We also carried out a metabolic study to explore whether the beneficial effects observed of 22SHC on glucose and lipid metabolism in vitro could be seen in vivo in rats. In the pharmacokinetic study, rats were given 50 mg/kg of [³H]22-S-hydroxycholesterol before absorption, distribution and excretion were monitored. In the metabolic study, the effect of 22SHC (30 mg/kg/day for 3 weeks) in rats on body weight gain [chow and high-fat diet (HFD)], serum lipids triacylglycerol (TAG) content and gene expression in liver and skeletal muscle were examined. Results showed that 22SHC was well absorbed after oral administration and distributed to most organs and mainly excreted in feces. Rats receiving 22SHC gained less body weight than their controls regardless whether the animals received chow diet or HFD. Moreover, we observed that animals receiving HFD had elevated levels of serum TAG while this was not observed for animals on HFD supplemented with 22SHC. The amount of TAG in liver was reduced after 22SHC treatment in animals receiving either chow diet or HFD. Gene expression analysis revealed that two genes (carnitine palmitoyltransferase 2 and uncoupling protein 3) involved in fatty acid oxidation and energy dissipation were increased in liver. Ucp3 expression (both protein and mRNA level) was increased in skeletal muscle, but insulin-stimulated glucose uptake and TAG content were unchanged. In conclusion, 22SHC seems to be an interesting model substance in the search of treatments for disorders involving aberrations in lipid metabolism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H9384 22(R)-Hydroxycholesterol, ≥98%
C27H46O2
H5884
22(S)-Hydroxycholesterol
C27H46O2