EMAIL THIS PAGE TO A FRIEND
European archives of oto-rhino-laryngology : official journal of the European Federation of Oto-Rhino-Laryngological Societies (EUFOS) : affiliated with the German Society for Oto-Rhino-Laryngology - Head and Neck Surgery
Preparation of nose for nasal endoscopy: cotton pledget packing versus topical spray. A prospective randomized blinded study.
PMID 22427029
Abstract
During nasal endoscopy it is essential to have proper visualization of structures with minimal discomfort to patient and surgeon. For this it is essential that the nose is well prepared before the procedure. The main objective of the study is to compare and evaluate the efficacy of cotton pledget packing versus topical sprays in preparation of nose for nasal endoscopy. The method includes prospective randomized blinded study on 100 patients. Patients were randomly divided in two groups. In first group the nose was packed with 4% lignocaine with xylometazoline nasal drops and in the other group it was prepared with 10% lignocaine topical spray and xylometazoline nose drops. Following the procedure, patient and the surgeon were asked a pre-formed questionnaire to know their experience during endoscopy. It was observed the packing group required more preparatory time as compared to the spray group. The group which was packed had less discomfort, less pain while endoscopy. The visualization of structures was significantly better in the packed group. Eight patients in the packed group did have some mucosal bleed during the process of packing which was not seen in the spray group. Both methods of preparation have merits and demerits but in terms of discomfort, pain during procedure and visualization of structure, packing of nasal cavity with 4% lignocaine and xylometazoline drops is better than spraying of nose with 10% lignocaine and xylometazoline drops.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
X6000
Xylometazoline hydrochloride
C16H24N2 · HCl