PloS one
A role for phospholipase D3 in myotube formation.
PMID 22428023
Abstract
Phospholipase D3 (PLD3) is a non-classical, poorly characterized member of the PLD superfamily of signaling enzymes. PLD3 is a type II glycoprotein associated with the endoplasmic reticulum, is expressed in a wide range of tissues and cells, and undergoes dramatic upregulation in neurons and muscle cells during differentiation. Using an in vitro skeletal muscle differentiation system, we define the ER-tethering mechanism and report that increased PLD3 expression enhances myotube formation, whereas a putatively dominant-negative PLD3 mutant isoform reduces myotube formation. ER stress, which also enhances myotube formation, is shown here to increase PLD3 expression levels. PLD3 protein was observed to localize to a restricted set of subcellular membrane sites in myotubes that may derive from or constitute a subdomain of the endoplasmic reticulum. These findings suggest that PLD3 plays a role in myogenesis during myotube formation, potentially in the events surrounding ER reorganization.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV44768
Anti-PLD3 antibody produced in rabbit, affinity isolated antibody