EMAIL THIS PAGE TO A FRIEND
Natural product communications
Complete NMR assignments of tubulosine.
PMID 22428233
Abstract
This article reports the structural elucidation of the Alangium alkaloid, tubulosine (1) on the basis of systematic 2D-NMR analyses (DEPT, COSY, TOCSY, NOESY, ROESY, HMQC and HMBC). The data obtained allowed the unambiguous assignment of all proton and carbon signals in 1 for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E2375
Emetine dihydrochloride hydrate
C29H40N2O4 · 2HCl · xH2O