EMAIL THIS PAGE TO A FRIEND
Natural product communications
GC-MS analysis of Ziziphora clinopodioides essential oil from North Xinjiang, China.
PMID 22428253
Abstract
The essential oils of Ziziphora clinopodioide Lam. from four different production areas (Banfang ditch; Altay mountains; Tuoli; Terks) were investigated. The oils were extracted by hydro-distillation and analyzed by gas chromatography-mass spectrometry (GC-MS). Seventeen constituents were identified in the essential oil from Banfang ditch, 20 in that from the Altay mountains, 12 in the Tuoli essential oil, and 9 in the Terks sample. The major components of the oils were pulegone (67.6%, 32.5%, 86.4%, and 82.1%) and p-menthanone (14.8%, 43.7%, 3.2%, and 8.2% from the Banfang ditch, Tuoli, Altay mountains, and Terks samples, respectively).