EMAIL THIS PAGE TO A FRIEND
Compendium of continuing education in dentistry (Jamesburg, N.J. : 1995)
SPRIX (ketorolac tromethamine) nasal spray: a novel nonopioid alternative for managing moderate to moderately severe dental pain.
PMID 22428363
Abstract
In summary, SPRIX is a nonopioid alternative for the management of moderate to moderately severe pain. SPRIX offers dentists, physicians, and patients a new non-opioid option to control acute moderate to moderately severe pain in situations in which use of an IM or IV access is not feasible or not wanted. SPRIX is a valuable treatment option for patients with nausea or vomiting, those unable to take oral medications, and those unable to tolerate the side effects of opioids. In ambulatory acute pain settings, use of SPRIX will allow patients who need to remain alert to receive effective pain control. Currently, there are no nonopioid alternatives for the treatment of moderate to moderately severe pain other than ketorolac. In patients with more severe pain states, the combination of opioids and SPRIX provides unique advantages in maximizing analgesia while minimizing the unwanted adverse effects of both classes of drugs (referred to as multimodal or "balanced analgesia").