EMAIL THIS PAGE TO A FRIEND
Journal of the science of food and agriculture
The effect of cocoa, soy, oats and fish oil on metabolic syndrome in rats.
PMID 22430394
Abstract
The effect of functional foods alone or in combination (cocoa + soy + oats + fish oil) on hepatic damage in rats affected with metabolic syndrome was investigated. Rats that were given cocoa showed a decrease in the levels of triglycerides (TGs) and glucose (63 and 32% respectively) as well as a decrease in blood pressure (15%). Animals fed with soy showed a reduction of 21% in total cholesterol, 15% in blood pressure and 44% in TGs, while feeding oats reduced the concentration of TGs by 53% (P < 0.5). Fish oil caused a reduction in TGs (56%) and glucose (26%). The effect on blood pressure was statistically significant for the groups supplemented with cocoa, soy, cocoa + oats and the total mix. The main finding was a reduction in liver steatosis in animals supplemented with cocoa + oats (from 30 to 4.7% steatosis). Cocoa or fish oil alone did not protect the liver from damage, while cocoa + fish oil did. The most relevant effects were that the cocoa + oats mix decreased steatosis by a very large percentage, as did the cocoa + fish oil mix and the mix of all four functional foods.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5157
Campesterol, ~65%
C28H48O