EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Tetravalent dysprosium in a perovskite-type oxide.
PMID 22430942
Abstract
The existence of tetravalent dysprosium in perovskite-type oxide barium zirconate is confirmed in this work. This discovery will stimulate many researchers in diverse fields such as catalysts, solid state ionics, sensors, and fluorescent materials.