EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Synthesis and characterization of amphiphilic reduced graphene oxide with epoxidized methyl oleate.
PMID 22431169
Abstract
Amphiphilic reduced graphene oxide is obtained by oleo-functionalization with epoxidized methyl oleate (renewable feedstock) using a green process. The excellent diverse solvent-dispersivity of the oleo-reduced amphiphilic graphene and its reduction chemistry are confirmed in this study. Oleo-reduction of amphiphilic graphene is amenable to industrially viable processes to produce future graphene-based polymer composites and systems.