EMAIL THIS PAGE TO A FRIEND
Journal of mass spectrometry : JMS
Mass spectrometry of metal complexes of bis-quinolizidine alkaloids: EI and ESI mass spectral study of Co(+2), Ni(+2), Cu(+2) and Zn(+2) 2-methylsparteine complexes.
PMID 22431461