EMAIL THIS PAGE TO A FRIEND
Current clinical pharmacology
Current status of CETP inhibitors in the treatment of hyperlipidemia: an update.
PMID 22432840
Abstract
The inverse relationship between HDL-C and cardiovascular disease risk suggests that increasing HDL-C could potentially reduce the disease risk. Reverse cholesterol transport is considered to be the primary mechanism by which HDL-C exerts its anti-atherogenic effects. A key regulator of RCT is cholesteryl ester transfer protein (CETP). Inhibition of CETP has been identified as a possible strategy for substantially increasing HDL-C levels and CETP inhibitors have demonstrated clinical efficacy in preliminary clinical trials. The development of this novel class suffered a major setback when the major phase 3 trial of torcetrapib, the first CETP inhibitior was prematurely terminated due to an increase in cardiovascular and noncardiovascular mortality. Subsequent animal and clinical studies have shown that the increase in cardiovascular mortality reported with torcetrapib was molecule specific and independent of its CETP inhibition effect. The other two CETP inhibitors i.e. dalcetrapib and anacetrapib were well tolerated in phase I and II clinical trials and unlike torcetrapib, did not affect blood pressure and aldosterone levels. In this review article the authors have discussed the lessons learned from torcetrapib failure and important preclinical and clinical developments of CETP inhibitors and their role in management of hyperlipidemia and cardiovascular risk reduction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PZ0170
Torcetrapib, ≥98% (HPLC)
C26H25F9N2O4