EMAIL THIS PAGE TO A FRIEND
Fetal and pediatric pathology
Aggressive sinonasal natural killer/T-cell lymphoma mimicking refractory sinusitis in a 4-year-old boy.
PMID 22432853
Abstract
Pediatric patients with nasal symptoms are common, and most of them usually have inflammatory diseases, such as sinusitis, chronic rhinitis, nasal polyp, or adenotonsillar hypertrophy. Rarely, however, these inflammatory symptoms may be associated with more sinister pathology. Recently, we experienced a case of a 4-year-old boy with sinonasal natural killer (NK)/T-cell lymphoma whose initial symptoms were of nasal obstruction and mucopurulent nasal discharge.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4292
Cefoperazone sodium salt, 870-1015 μg/mg (anhydrous basis)
C25H26N9NaO8S2