EMAIL THIS PAGE TO A FRIEND
Internal medicine journal
Carcinoid crisis induced by repeated abdominal examination.
PMID 22432991
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0132
Lanreotide acetate, ≥98% (HPLC)
C54H69N11O10S2 · xC2H4O2