EMAIL THIS PAGE TO A FRIEND
Ambio
Characterization of photovoltaic performance of the dye-sensitized solar cell with a novel ruthenium complex having a bisdemethoxycurcumin as a ligand.
PMID 22434443
Abstract
The first example of a ruthenium sensitizer (TUS-22) having a natural dye, bisdemethoxycurcumin, as a ligand has been synthesized. The dye-sensitized solar cell based on this novel dye showed 5.8% conversion efficiency under AM 1.5 (100 mW/cm(2)) irradiation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1075305 Bisdesmethoxycurcumin, United States Pharmacopeia (USP) Reference Standard
C19H16O4