EMAIL THIS PAGE TO A FRIEND
Immunological reviews
IKK biology.
PMID 22435559
Abstract
The inhibitor of nuclear factor-κB (IκB) kinase (IKK) complex is the master regulator of the NF-κB signaling pathway. The activation of the IKK complex is a tightly regulated, highly stimulus-specific, and target-specific event that is essential for the plethora of functions attributed to NF-κB. More recently, NF-κB-independent roles of IKK members have brought increased complexity to its biological function. This review highlights some of the major advances in the studies of the process of IKK activation and the biological roles of IKK family members, with a focus on NF-κB-independent functions. Understanding these complex processes is essential for targeting IKK for therapeutics.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV30005
Anti-IKBKG antibody produced in rabbit, IgG fraction of antiserum