EMAIL THIS PAGE TO A FRIEND
British journal of clinical pharmacology
Suicide attempt with an overdose of imatinib.
PMID 22435728
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T-910
Triazolam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material
C17H12Cl2N4