EMAIL THIS PAGE TO A FRIEND
Colloids and surfaces. B, Biointerfaces
Effect of aggregation behavior of gelatin in aqueous solution on the grafting density of gelatin modified with glycidol.
PMID 22436469
Abstract
The effect of aggregation behavior of gelatin in aqueous solution on the grafting density of glycidol grafted gelatin polymers (GGG polymers) was investigated. The grafting density was measured using the Van Slyke method by calculating the conversion rate of free - NH(2) groups of gelatin. The conversion rate reached peak values at 6% and 14% of the gelatin aqueous solution. SEM micrographs displayed a series of structural transitions (i.e., spherical, spindle, butterfly, irregular and dendritic aggregates) at varying concentrations from 2% to 16% (w/w) at an interval of 2% (w/w). The spindle aggregates reappeared at the concentrations of 6% and 14%. Viscosity measurements indicated that the physicochemical properties of the gelatin solution had changed with increasing concentration. UV and CD analysis indicated that hydrophobic interactions competed with hydrogen bonding, and the random coils partly transformed to β-sheet structure by changing the concentration. Zeta potential and pH data confirmed the increasing electrostatic repulsion associated with increasing the hydrophobic region. XPS analysis revealed that the elemental composition of the gelatin particle surface changed with variation in the aggregate structure, determining the monotonic variation of the grafting density with increasing concentration. Results demonstrate that aggregation behavior of gelatin in aqueous solution plays a crucial role in deciding the grafting density of gelatin modified products.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
480819
(R)-(+)-Glycidol, 97%, optical purity ee: 98% (GLC)
C3H6O2
G5809
Glycidol, 96%
C3H6O2