EMAIL THIS PAGE TO A FRIEND
Gynecological endocrinology : the official journal of the International Society of Gynecological Endocrinology
Outcomes from ovarian hyperstimulation following the sole administration of gonadotrophin-releasing hormone agonist in the context of in vitro fertilization: report of two cases and review of the literature.
PMID 22439899
Abstract
To report the outcomes from two cases of ovarian stimulation following the sole administration of gonadotrophin-releasing hormone agonist (GnRHa) in the context of in vitro fertilization (IVF). A case study was conducted. National Referral Unit of Reproductive Medicine. Two infertile women undergoing IVF participated in the study. Controlled ovarian hyperstimulation using a long protocol. GnRHa (Buserelin) was started in the luteal phase, in a dose of 600 µg/day, for 12 days. Number of retrieved oocytes, fertilization rate, number of embryos transferred, implantation rate, ongoing pregnancy, and live birth. Both women underwent egg retrieval and transfer of good quality embryos. One of them conceived and recently gave birth to a healthy full-term baby. The ovarian hyperstimulation after the sole administration of GnRHa is a rare condition. Oocyte retrieval may be a reasonable treatment under these circumstances instead of cycle cancellation. As far as it is known, this is the third case reported of a live birth following the sole administration of GnRHa in the context of IVF.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B3303 Buserelin acetate salt, ≥90% (HPLC)
C60H86N16O13 · C2H4O2