EMAIL THIS PAGE TO A FRIEND
ChemSusChem
Electrochemical synthesis of adiponitrile from the renewable raw material glutamic acid.
PMID 22441826
Abstract
Current affairs: Adiponitrile, used to produce nylon 6.6, is prepared from the renewable compound glutamic acid by an electrochemical route, involving electro-oxidative decarboxylation and Kolbe coupling reactions. The new route is an example of the use of glutamic acid as a versatile substrate in the transformation of biomass into chemicals. Also, it highlights the use of electrochemical methods in biomass conversion.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D77001
Adiponitrile, 99%
C6H8N2