EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Semiconducting polymers prepared by direct arylation polycondensation.
PMID 22441886
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
773859
5-Octylthieno[3,4-c]pyrrole-4,6-dione, 98%
C14H19NO2S