EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
An efficient flow-photochemical synthesis of 5H-furanones leads to an understanding of torquoselectivity in cyclobutenone rearrangements.
PMID 22441937