EMAIL THIS PAGE TO A FRIEND
The Journal of Biological Chemistry
The AT-hook motif-containing protein AHL22 regulates flowering initiation by modifying FLOWERING LOCUS T chromatin in Arabidopsis.
PMID 22442143