EMAIL THIS PAGE TO A FRIEND
Traffic (Copenhagen, Denmark)
SNAP-tag based proteomics approach for the study of the retrograde route.
PMID 22443104
Abstract
Proteomics is a powerful technique for protein identification at large scales. A number of proteomics approaches have been developed to study the steady state composition of intracellular compartments. Here, we report a novel vectorial proteomics strategy to identify plasma membrane proteins that undergo retrograde transport to the trans-Golgi network (TGN). This strategy is based on the covalent modification of the plasma membrane proteome with a membrane impermeable benzylguanine derivative. Benzylguanine-tagged plasma membrane proteins that are subsequently targeted to the retrograde route are covalently captured by a TGN-localized SNAP-tagged fusion protein, which allows for their identification. The approach was validated step-by-step using a well explored retrograde cargo protein, the B-subunit of Shiga toxin. It was then extended to the proteomics format. Among other hits we found one of the historically first identified cargo proteins that undergo retrograde transport, which further validated our approach. Most of the other hits were kinases, receptors or transporters. In conclusion, we have pioneered a vectorial proteomics approach that complements traditional methods for the study of retrograde protein trafficking. This approach is of generic nature and could in principle be extended to other endocytic pathways.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B2292
O6-Benzylguanine, ≥98% (TLC), solid
C12H11N5O