EMAIL THIS PAGE TO A FRIEND
Tandlaegebladet
[Lead foil artifacts as a result of unintentional bending of dental X-ray films].
PMID 2244316
Abstract
The lead foil of dental x-ray film packets may cause a number of artifacts; especially well known are the "tractor treads" and "herringbone patterns" following a backward filmpositioning in the patient's mouth. In the present paper a specific and seldomly occurring lead foil artifact is described which is caused by bending of the film packet. Four cases collected over a 5-year period are presented. Although different the four cases show a number of similarities. They appear as radiopaque areas covering the upper or lower halves of the films. Their borders show the characteristic "sandbar-pattern" previously described by the author. The explanation of their origin is that the patient has been occluding on a part of the film packet which has been bent to a nearly horizontal position. The x-rays directed against the edge of the lead foil will be absorbed causing irregular radiopaque areas on the film lined by "sandbar-pattern". When the film packet is bent, the lead foil will be stretched and drawn a little away from the edges of the films. Consequently the x-rays will be able to expose the edges of the films. Irregularities of the foil will give rise to concentric "bars". The lead foil is only of limited radiopacity, but due to the oblique direction of the x-rays through the foil the absorbing effect is intensified.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF04438919
Lead, foil, 50x50mm, thickness 0.25mm, as rolled, 99.95%
Pb
GF21166321
Lead, foil, 100x100mm, thickness 3.0mm, as rolled, 99.95%
Pb
GF24521329
Lead, foil, 100x100mm, thickness 3.0mm, as rolled, 99.99+%
Pb
GF97211572
Lead, foil, 100x100mm, thickness 6.0mm, as rolled, 99.99+%
Pb
GF26692263
Lead, foil, 150x150mm, thickness 0.125mm, as rolled, 99.95%
Pb
GF86984229
Lead, foil, 150x150mm, thickness 0.15mm, as rolled, 99.95%
Pb
GF22298777
Lead, foil, 150x150mm, thickness 0.1mm, as rolled, 99.95%
Pb
GF58005301
Lead, foil, 150x150mm, thickness 0.25mm, as rolled, 99.95%
Pb
GF21793372
Lead, foil, 150x150mm, thickness 0.55mm, as rolled, 99.999%
Pb
GF96512597
Lead, foil, 150x150mm, thickness 0.5mm, as rolled, 99.95%
Pb
GF74259793
Lead, foil, 150x150mm, thickness 1.0mm, as rolled, 99.5%
Pb
GF57430638
Lead, foil, 150x150mm, thickness 1.0mm, as rolled, 99.95%
Pb
GF60047760
Lead, foil, 250x250mm, thickness 3.0mm, as rolled, 99.95%
Pb
GF05521160
Lead, foil, 25x25mm, thickness 0.125mm, as rolled, 99.95%
Pb
GF42741637
Lead, foil, 25x25mm, thickness 0.125mm, as rolled, 99.99+%
Pb
GF60380940
Lead, foil, 25x25mm, thickness 0.15mm, as rolled, 99.95%
Pb
GF85562994
Lead, foil, 25x25mm, thickness 0.1mm, as rolled, 99.95%
Pb
GF74435519
Lead, foil, 25x25mm, thickness 0.25mm, as rolled, 99.95%
Pb
GF10987757
Lead, foil, 25x25mm, thickness 0.25mm, as rolled, 99.99+%
Pb
GF03985724
Lead, foil, 25x25mm, thickness 0.55mm, as rolled, 99.999%
Pb
GF73081852
Lead, foil, 25x25mm, thickness 0.5mm, as rolled, 99.95%
Pb
GF68898188
Lead, foil, 25x25mm, thickness 0.5mm, as rolled, 99.99+%
Pb
GF90025129
Lead, foil, 25x25mm, thickness 1.0mm, as rolled, 99.5%
Pb
GF55764041
Lead, foil, 25x25mm, thickness 1.0mm, as rolled, 99.95%
Pb
GF85349346
Lead, foil, 25x25mm, thickness 1.0mm, as rolled, 99.99+%
Pb
GF70374810
Lead, foil, 25x25mm, thickness 1.5mm, as rolled, 99.99+%
Pb
GF22900766
Lead, foil, 25x25mm, thickness 3.0mm, as rolled, 99.99+%
Pb
GF36533505
Lead, foil, 300x300mm, thickness 0.125mm, as rolled, 99.95%
Pb
GF00434843
Lead, foil, 300x300mm, thickness 0.15mm, as rolled, 99.95%
Pb
GF87346806
Lead, foil, 300x300mm, thickness 0.1mm, as rolled, 99.95%
Pb