EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Design and synthesis of novel opioid ligands with an azabicyclo[2.2.2]octane skeleton and their pharmacologies.
PMID 22445285
Abstract
A novel opioid ligand possessing a stable and cyclic imine 16 and its derivatives with an azabicyclo[2.2.2]octane skeleton were synthesized. The imine 16 showed higher affinity for the μ receptor than compound 21 with an oxabicyclo[2.2.2]octane skeleton. Azabicyclo[2.2.2]octane derivative 18d with a cyclopropylmethyl group on the 8-nitrogen showed the highest affinity for the μ receptor among the synthesized derivatives.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74821
Octane, puriss. p.a., ≥99.0% (GC)
C8H18
412236
Octane, reagent grade, 98%
C8H18