EMAIL THIS PAGE TO A FRIEND
Molecular bioSystems
Dynamic eicosanoid responses upon different inhibitor and combination treatments on the arachidonic acid metabolic network.
PMID 22446875
Abstract
The arachidonic acid (AA) metabolic network produces key inflammatory mediators which have been considered as hallmark contributors in various inflammatory related diseases. Enzymes in this network, such as 5-lipoxygenase (5-LOX), cyclooxygenase (COX), leukotriene A(4) hydrolase (LTA4H) and prostaglandin E synthase (PGES), have been used as targets for anti-inflammatory drug discovery. Multi-target drugs and drug combinations have also been developed for this network. However, how the inhibitors alter the dynamics of metabolite production and which combinatorial target intervention solutions are better needs further exploration. We did a system based intervention analysis on the AA metabolic network. Using an LC-MS/MS method, we quantitatively studied the eicosanoid metabolites responses of AA metabolic network during stimulation of Sprague Dawley rat blood samples with the calcium ionophore. Our results indicate that inhibiting the upstream rather than the downstream target of 5-LOX pathway will simultaneously alter the AA metabolism to the COX pathway (and vice versa). Therefore, single-target inhibitors cannot control all the inflammatory mediators at the same time. We also suggest that in the case of multiple-target anti-inflammatory solutions, the combination of inhibitors of the downstream enzymes may have stronger inhibition efficiency and cause less side-effects compared to the other solutions. One therapeutic strategy, LTA4H/COX inhibition solution, was found promising for the intervention of inflammatory mediator biosynthesis and at the same time stimulating the production of anti-inflammatory agents.
Related Materials