EMAIL THIS PAGE TO A FRIEND
Advanced materials (Deerfield Beach, Fla.)
Nanocomposite "superhighways" by solution assembly of semiconductor nanostructures with ligand-functionalized conjugated polymers.
PMID 22451039
Abstract
Poly(3-hexyl thiophene) containing chain-end thiols or phosphonic acids is crystallized to yield nanowires with the functional groups at the wire edges. CdSe quantum dots and nanorods associate with these fibrils, leading to 'superhighways' that consist of alternating parallel lanes of conjugated polymer and CdSe.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
215112
Phosphorous acid, 99%
H3O3P
04115
Phosphorous acid solution, ≥50%
H3O3P