EMAIL THIS PAGE TO A FRIEND
Organic letters
Regioselective Cu(I)-catalyzed tandem A3-coupling/decarboxylative coupling to 3-amino-1,4-enynes.
PMID 22455453
Abstract
An efficient and novel copper-mediated protocol for the synthesis of 3-amino-1,4-enynes from glyoxylic acid, an amine, and an alkyne was developed. This new reaction involving two sequential C-C bond formations is air and moisture tolerant and proceeds via a tandem A(3)-coupling and a selective decarboxylative coupling.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
260150
Glyoxylic acid solution, 50 wt. % in H2O
C2H2O3