Canadian journal of microbiology
Purification and characterization of azurin from the methylamine-utilizing obligate methylotroph Methylobacillus flagellatus KT.
PMID 22455796
Methylamine dehydrogenase (MADH) and azurin were purified from the periplasmic fraction of the methylamine-grown obligate methylotroph Methylobacillus flagellatus KT. The molecular mass of the purified azurin was 16.3 kDa, as measured by SDS-PAGE, or 13 920 Da as determined by MALDI-TOF mass spectrometry. Azurin of M. flagellatus KT contained 1 copper atom per molecule and had an absorption maximum at 620 nm in the oxidized state. The redox potential of azurin measured at pH 7.0 by square-wave voltammetry was +275 mV versus normal hydrogen electrode. MADH reduced azurin in the presence of methylamine, indicating that this cupredoxin is likely to be the physiological electron acceptor for MADH in the electron transport chain of the methylotroph. A scheme of electron transport functioning in M. flagellatus KТ during methylamine oxidation is proposed.
Related Materials