EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
One-pot synthesis of 4-substituted isoquinolinium zwitterionic salts by metal-free C-H bond activation.
PMID 22456497
Abstract
The unprecedented isoquinolinium zwitterionic salts with an unusual C-4 substitution pattern were efficiently prepared via the multicomponent reaction of in situ formed N-benzylisoquinolinium bromide with aromatic aldehydes and cyclic 1,3-dicarbonyl compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I28208
Isoquinoline, 97%
C9H7N
282162
Isoquinoline, technical grade, 90-92%
C9H7N