EMAIL THIS PAGE TO A FRIEND
Molecular biology of the cell
Protein kinase C δ regulates the release of collagen type I from vascular smooth muscle cells via regulation of Cdc42.
PMID 22456512
Abstract
Collagen type I is the most abundant component of extracellular matrix in the arterial wall. Mice knocked out for the protein kinase C δ gene (PKCδ KO) show a marked reduction of collagen I in the arterial wall. The lack of PKCδ diminished the ability of arterial smooth muscle cells (SMCs) to secrete collagen I without significantly altering the intracellular collagen content. Moreover, the unsecreted collagen I molecules accumulate in large perinuclear puncta. These perinuclear structures colocalize with the trans-Golgi network (TGN) marker TGN38 and to a lesser degree with cis-Golgi marker (GM130) but not with early endosomal marker (EEA1). Associated with diminished collagen I secretion, PKCδ KO SMCs exhibit a significant reduction in levels of cell division cycle 42 (Cdc42) protein and mRNA. Restoring PKCδ expression partially rescues Cdc42 expression and collagen I secretion in PKCδ KO SMCs. Inhibition of Cdc42 expression or activity with small interfering RNA or secramine A in PKCδ WT SMCs eliminates collagen I secretion. Conversely, restoring Cdc42 expression in PKCδ KO SMCs enables collagen I secretion. Taken together, our data demonstrate that PKCδ mediates collagen I secretion from SMCs, likely through a Cdc42-dependent mechanism.