EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Palladium-catalyzed N-acylation of monosubstituted ureas using near-stoichiometric carbon monoxide.
PMID 22458554
Abstract
The palladium-catalyzed carbonylation of urea derivatives with aryl iodides and bromides afforded N-benzoyl ureas (20 examples) in yields attaining quantitative via the application of near-stoichiometric amounts of carbon monoxide generated from the decarbonylation of the CO precursor, 9-methylfluorene-9-carbonyl chloride. The synthetic protocol displayed good functional group tolerance. The methodology is also highly suitable for (13)C isotope labeling, which was demonstrated through the synthesis of three benzoyl ureas, including the insecticide triflumuron, whereby (13)CO was incorporated into the core structure.