EMAIL THIS PAGE TO A FRIEND
Profiles of drug substances, excipients, and related methodology
Flurbiprofen.
PMID 22469318
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F0285200
Flurbiprofen, European Pharmacopoeia (EP) Reference Standard
C15H13FO2